siRNA Details
Gene Symbol: WDR38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045476.2 | NP_001038941.1 | 5 | 537 | ||
NM_001276374.1 | NP_001263303.1 | 5 | 537 | |||
NM_001276375.1 | NP_001263304.1 | 5 | 504 | |||
NM_001276376.1 | NP_001263305.1 | 4 | 484 | |||
GenBank mRNA | BC127949.1 | - | 5 | 501 | 40132852 | |
BC150645.1 | - | 5 | 537 | 9057015 | ||
BC171828.1 | - | 5 | 537 | 9054482 | ||
BC171830.1 | - | 5 | 504 | 9054484 | ||
BC171837.1 | - | 4 | 484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
