siRNA Details
Gene Symbol: IGFBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007563.2 | NP_001007564.1 | 5 | 1629 | ||
GenBank mRNA | AK055716.1 | - | 1 | 100 | ||
AK131521.1 | - | 5 | 1182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
