siRNA Details
Gene Symbol: TMEM63B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318792.1 | NP_001305721.1 | 12 | 1106 | ||
NM_018426.2 | NP_060896.1 | 12 | 1072 | |||
XM_005249211.3 | XP_005249268.1 | 14 | 1452 | |||
XM_005249213.4 | XP_005249270.1 | 13 | 1379 | |||
XM_005249214.4 | XP_005249271.1 | 11 | 966 | |||
XM_005249215.4 | XP_005249272.1 | 12 | 1271 | |||
XM_005249217.1 | XP_005249274.1 | 12 | 1144 | |||
XM_006715135.1 | XP_006715198.1 | 12 | 1128 | |||
XM_017010999.1 | XP_016866488.1 | 14 | 1329 | |||
XM_017011000.1 | XP_016866489.1 | 13 | 1310 | |||
XM_017011001.1 | XP_016866490.1 | 11 | 1010 | |||
XM_017011002.1 | XP_016866491.1 | 11 | 1113 | |||
GenBank mRNA | AK093629.1 | - | 12 | 1128 | ||
AL834238.1 | - | 5 | 370 | |||
BC022095.1 | - | 9 | 709 | |||
BC136769.1 | - | 11 | 948 | 9020759 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
