siRNA Details
Gene Symbol: KIAA0319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168374.1 | NP_001161846.1 | 4 | 1039 | ||
NM_001168375.1 | NP_001161847.1 | 3 | 923 | |||
NM_001168376.1 | NP_001161848.1 | 2 | 763 | |||
NM_001168377.1 | NP_001161849.1 | 3 | 948 | |||
NM_014809.3 | NP_055624.2 | 3 | 948 | |||
XM_011515025.2 | XP_011513327.1 | 3 | 705 | |||
XM_011515026.2 | XP_011513328.1 | 2 | 666 | |||
XM_017011541.1 | XP_016867030.1 | 4 | 1019 | |||
XM_017011542.1 | XP_016867031.1 | 3 | 666 | |||
XM_017011543.1 | XP_016867032.1 | 3 | 666 | |||
XM_017011544.1 | XP_016867033.1 | 3 | 739 | |||
XM_017011545.1 | XP_016867034.1 | 3 | 661 | |||
XM_017011546.1 | XP_016867035.1 | 3 | 666 | |||
XM_017011547.1 | XP_016867036.1 | 3 | 666 | |||
XM_017011548.1 | XP_016867037.1 | 3 | 739 | |||
XM_017011549.1 | XP_016867038.1 | 3 | 696 | |||
XM_017011550.1 | XP_016867039.1 | 3 | 666 | |||
XM_017011551.1 | XP_016867040.1 | 3 | 666 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
