siRNA Details
Gene Symbol: MDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014641.2 | NP_055456.2 | 13 | 6378 | ||
XM_005249492.2 | XP_005249549.1 | 13 | 6282 | |||
XM_005249493.3 | XP_005249550.1 | 13 | 6279 | |||
XM_005249494.4 | XP_005249551.1 | 13 | 6681 | |||
XM_005249497.3 | XP_005249554.1 | 13 | 5712 | |||
XM_005249498.3 | XP_005249555.1 | 12 | 5490 | |||
XM_011515001.2 | XP_011513303.1 | 13 | 6109 | |||
XM_011515003.2 | XP_011513305.1 | 13 | 5964 | |||
XM_011515004.2 | XP_011513306.1 | 14 | 5993 | |||
XM_017011519.1 | XP_016867008.1 | 13 | 6061 | |||
XM_017011520.1 | XP_016867009.1 | 13 | 5977 | |||
XM_017011521.1 | XP_016867010.1 | 13 | 5410 | |||
XM_017011522.1 | XP_016867011.1 | 12 | 5487 | |||
GenBank mRNA | AK302434.1 | - | 14 | 5066 | ||
BC110645.1 | - | 8 | 2035 | |||
BC131491.1 | - | 9 | 3349 | |||
BC152556.1 | - | 13 | 5090 | 9007258 | ||
CR749828.1 | - | 9 | 4010 | |||
D79992.1 | - | 12 | 5951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
