siRNA Details
Gene Symbol: ZFP57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109809.2 | NP_001103279.2 | 3 | 426 | ||
XM_006715087.3 | XP_006715150.1 | 3 | 241 | |||
XM_011514570.2 | XP_011512872.1 | 4 | 727 | |||
GenBank mRNA | BC157878.1 | - | 3 | 426 | 8862701 | |
BC171888.1 | - | 3 | 426 | 9054542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
