siRNA Details
Gene Symbol: GTF2H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001517.4 | NP_001508.1 | 10 | 1124 | ||
GenBank mRNA | AK222607.1 | - | 10 | 1028 | ||
AK298058.1 | - | 10 | 1172 | |||
BC004935.1 | - | 10 | 1096 | 3606718 | ||
BC016302.2 | - | 10 | 1096 | 3830902 | ||
BT007321.1 | - | 9 | 917 | |||
Y07595.1 | - | 10 | 1044 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
