siRNA Details
Gene Symbol: C6orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145020.1 | NP_001138492.1 | 1 | 61 | ||
NR_026737.1 | 1 | 61 | ||||
XM_006715088.3 | XP_006715151.1 | 1 | 230 | |||
XM_011514575.2 | XP_011512877.1 | 1 | 230 | |||
GenBank mRNA | BC016820.1 | - | 1 | 65 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
