siRNA Details
Gene Symbol: FDXACB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138378.2 | NP_612387.1 | 5 | 1677 | ||
NR_038364.1 | 4 | 1520 | ||||
GenBank mRNA | AK097483.1 | - | 4 | 1508 | ||
AK300795.1 | - | 4 | 1151 | |||
BC006136.1 | - | 5 | 1356 | |||
BC127680.1 | - | 5 | 1400 | 40127680 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
