siRNA Details
Gene Symbol: UNC13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080534.1 | NP_001074003.1 | 17 | 4830 | ||
XM_005254394.4 | XP_005254451.1 | 18 | 5781 | |||
XM_017022220.1 | XP_016877709.1 | 22 | 5701 | |||
XM_017022221.1 | XP_016877710.1 | 21 | 5581 | |||
XM_017022222.1 | XP_016877711.1 | 20 | 5479 | |||
XM_017022223.1 | XP_016877712.1 | 18 | 11007 | |||
XM_017022224.1 | XP_016877713.1 | 17 | 5707 | |||
XM_017022225.1 | XP_016877714.1 | 15 | 3132 | |||
GenBank mRNA | AK091491.1 | - | 14 | 1806 | ||
AK293996.1 | - | 7 | 747 | |||
AL834407.1 | - | 1 | 105 | |||
BC039494.1 | - | 10 | 1405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
