siRNA Details
Gene Symbol: MAPKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128608.1 | NP_001122080.1 | 32 | 5242 | ||
NM_001265611.1 | NP_001252540.1 | 30 | 4393 | |||
NM_014994.2 | NP_055809.2 | 31 | 5224 | |||
NR_049761.1 | 30 | 5034 | ||||
NR_049762.1 | 29 | 4985 | ||||
XM_006720438.1 | XP_006720501.1 | 30 | 5041 | |||
XM_006720439.2 | XP_006720502.1 | 18 | 3316 | |||
XM_011521383.1 | XP_011519685.1 | 31 | 5059 | |||
XM_017022017.1 | XP_016877506.1 | 30 | 5335 | |||
GenBank mRNA | AB011168.2 | - | 32 | 5152 | ||
AB177850.1 | - | 23 | 4066 | |||
AL833267.1 | - | 31 | 5144 | |||
BC113983.1 | - | 30 | 4285 | 40083186 | ||
BC114493.1 | - | 32 | 5134 | 40083182 | ||
BX640787.1 | - | 27 | 5602 | |||
BX648786.1 | - | 31 | 5175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
