siRNA Details
Gene Symbol: SULT6B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032377.1 | NP_001027549.1 | 1 | 86 | ||
GenBank mRNA | BC140797.1 | - | 1 | 86 | 9021665 | |
BC144573.1 | - | 1 | 86 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
