siRNA Details
Gene Symbol: HECW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020760.2 | NP_065811.1 | 2 | 421 | ||
XM_006712646.3 | XP_006712709.1 | 2 | 458 | |||
XM_017004563.1 | XP_016860052.1 | 2 | 458 | |||
GenBank mRNA | AB037722.1 | - | 2 | 421 | ||
BC117194.1 | - | 2 | 337 | 40125745 | ||
BC117198.1 | - | 2 | 337 | 40125749 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
