siRNA Details
Gene Symbol: MON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278469.1 | NP_001265398.1 | 33 | 5174 | ||
NM_001278470.1 | NP_001265399.1 | 32 | 5156 | |||
NM_001278471.1 | NP_001265400.1 | 32 | 5087 | |||
NM_001278472.1 | NP_001265401.1 | 34 | 5252 | |||
NM_015026.2 | NP_055841.2 | 33 | 5174 | |||
XM_017019041.1 | XP_016874530.1 | 34 | 5177 | |||
XM_017019042.1 | XP_016874531.1 | 33 | 5159 | |||
XM_017019043.1 | XP_016874532.1 | 35 | 5251 | |||
XM_017019044.1 | XP_016874533.1 | 35 | 5253 | |||
XM_017019045.1 | XP_016874534.1 | 36 | 4921 | |||
XM_017019046.1 | XP_016874535.1 | 30 | 4346 | |||
GenBank mRNA | AB017814.1 | - | 33 | 4899 | ||
AB028963.3 | - | 34 | 5145 | |||
AK022671.1 | - | 7 | 1351 | |||
AK092646.1 | - | 7 | 1938 | |||
AL833066.2 | - | 33 | 5169 | |||
AL834320.1 | - | 27 | 4132 | |||
BC136621.1 | - | 32 | 5125 | 9020611 | ||
BC144309.1 | - | 32 | 5056 | 9052829 | ||
BC151241.1 | - | 34 | 5168 | 8860514 | ||
BX537415.1 | - | 33 | 5172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
