siRNA Details
Gene Symbol: MON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278469.1 | NP_001265398.1 | 34 | 5348 | ||
NM_001278470.1 | NP_001265399.1 | 33 | 5330 | |||
NM_001278471.1 | NP_001265400.1 | 33 | 5261 | |||
NM_001278472.1 | NP_001265401.1 | 35 | 5426 | |||
NM_015026.2 | NP_055841.2 | 34 | 5348 | |||
XM_017019041.1 | XP_016874530.1 | 35 | 5351 | |||
XM_017019042.1 | XP_016874531.1 | 34 | 5333 | |||
XM_017019043.1 | XP_016874532.1 | 36 | 5425 | |||
XM_017019044.1 | XP_016874533.1 | 36 | 5427 | |||
XM_017019045.1 | XP_016874534.1 | 37 | 5095 | |||
XM_017019046.1 | XP_016874535.1 | 31 | 4520 | |||
GenBank mRNA | AB017814.1 | - | 34 | 5073 | ||
AB028963.3 | - | 35 | 5319 | |||
AK022671.1 | - | 8 | 1525 | |||
AK092646.1 | - | 8 | 2112 | |||
AL833066.2 | - | 34 | 5343 | |||
AL834320.1 | - | 28 | 4306 | |||
BC136621.1 | - | 33 | 5299 | 9020611 | ||
BC144309.1 | - | 33 | 5230 | 9052829 | ||
BC151241.1 | - | 35 | 5342 | 8860514 | ||
BX537415.1 | - | 34 | 5346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
