siRNA Details
Gene Symbol: FAM189A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015307.1 | NP_056122.1 | 11 | 1967 | ||
XM_011521407.2 | XP_011519709.1 | 12 | 2124 | |||
GenBank mRNA | AB011146.1 | - | 8 | 1567 | ||
AK295622.1 | - | 3 | 961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
