siRNA Details
Gene Symbol: PRDM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136239.1 | NP_001129711.1 | 8 | 2905 | ||
XM_011543726.2 | XP_011542028.1 | 7 | 2968 | |||
GenBank mRNA | AF272898.1 | - | 8 | 2555 | ||
AF272899.1 | - | 6 | 2302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
