siRNA Details
Gene Symbol: USP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015276.1 | NP_056091.1 | 13 | 4949 | ||
XM_005256575.2 | XP_005256632.1 | 13 | 4663 | |||
GenBank mRNA | AB028986.1 | - | 13 | 4956 | ||
AK025331.1 | - | 1 | 2472 | |||
AL049258.1 | - | 1 | 1905 | |||
AL110213.1 | - | 1 | 1225 | |||
BC007196.1 | - | 6 | 3690 | |||
BC013364.2 | - | 1 | 2509 | |||
BX640815.1 | - | 12 | 4532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
