siRNA Details
Gene Symbol: SMCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015295.2 | NP_056110.2 | 30 | 4107 | ||
XM_011525642.1 | XP_011523944.1 | 30 | 4107 | |||
XM_011525643.2 | XP_011523945.1 | 30 | 4107 | |||
XM_011525644.2 | XP_011523946.1 | 29 | 4015 | |||
XM_017025684.1 | XP_016881173.1 | 31 | 4285 | |||
GenBank mRNA | AB014550.1 | - | 4 | 449 | ||
AK126324.1 | - | 15 | 1738 | |||
AL080138.1 | - | 1 | 87 | |||
CR627458.1 | - | 19 | 2331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
