siRNA Details
Gene Symbol: SMCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015295.2 | NP_056110.2 | 33 | 4419 | ||
XM_011525642.1 | XP_011523944.1 | 33 | 4419 | |||
XM_011525643.2 | XP_011523945.1 | 33 | 4419 | |||
XM_011525644.2 | XP_011523946.1 | 32 | 4327 | |||
XM_017025684.1 | XP_016881173.1 | 34 | 4597 | |||
GenBank mRNA | AB014550.1 | - | 7 | 761 | ||
AK126324.1 | - | 18 | 2050 | |||
AL080138.1 | - | 4 | 399 | |||
CR627458.1 | - | 22 | 2643 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
