siRNA Details
Gene Symbol: ZMYM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190964.2 | NP_001177893.1 | 4 | 274 | ||
NM_001190965.2 | NP_001177894.1 | 3 | 289 | |||
NM_003453.4 | NP_003444.1 | 4 | 382 | |||
NM_197968.3 | NP_932072.1 | 3 | 335 | |||
XM_005266520.3 | XP_005266577.1 | 3 | 294 | |||
XM_011535216.2 | XP_011533518.1 | 3 | 225 | |||
XM_011535222.1 | XP_011533524.1 | 3 | 225 | |||
XM_017020728.1 | XP_016876217.1 | 3 | 225 | |||
XM_017020729.1 | XP_016876218.1 | 3 | 200 | |||
XM_017020731.1 | XP_016876220.1 | 3 | 278 | |||
XM_017020732.1 | XP_016876221.1 | 3 | 300 | |||
XM_017020733.1 | XP_016876222.1 | 4 | 327 | |||
XM_017020734.1 | XP_016876223.1 | 4 | 313 | |||
XM_017020735.1 | XP_016876224.1 | 3 | 277 | |||
XM_017020736.1 | XP_016876225.1 | 3 | 449 | |||
XM_017020737.1 | XP_016876226.1 | 3 | 224 | |||
XM_017020738.1 | XP_016876227.1 | 3 | 278 | |||
XM_017020739.1 | XP_016876228.1 | 3 | 225 | |||
GenBank mRNA | AJ224901.1 | - | 4 | 268 | ||
AK226118.1 | - | 3 | 272 | |||
AK289741.1 | - | 3 | 260 | |||
AK310505.1 | - | 3 | 324 | |||
AL136621.1 | - | 3 | 329 | |||
BC036372.1 | - | 3 | 335 | 4823008 | ||
BI460760.1 | - | 3 | 337 | |||
BX643074.1 | - | 3 | 284 | |||
BX647944.1 | - | 3 | 289 | |||
Y13472.1 | - | 4 | 274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
