siRNA Details
Gene Symbol: TBC1D27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025506.1 | XP_016880995.1 | 5 | 772 | ||
GenBank mRNA | AK024458.1 | - | 14 | 3662 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
