siRNA Details
Gene Symbol: FBRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142641.1 | NP_001136113.1 | 17 | 3619 | ||
XM_005266171.3 | XP_005266228.1 | 20 | 3850 | |||
XM_005266173.3 | XP_005266230.1 | 20 | 3845 | |||
XM_005266175.3 | XP_005266232.1 | 19 | 3784 | |||
XM_005266176.3 | XP_005266233.1 | 19 | 3726 | |||
XM_005266177.3 | XP_005266234.1 | 20 | 3719 | |||
XM_011534803.2 | XP_011533105.1 | 20 | 3909 | |||
XM_011534804.2 | XP_011533106.1 | 20 | 3906 | |||
XM_011534805.2 | XP_011533107.1 | 19 | 3840 | |||
XM_011534806.2 | XP_011533108.1 | 19 | 3843 | |||
XM_011534807.2 | XP_011533109.1 | 19 | 3785 | |||
XM_011534808.2 | XP_011533110.1 | 20 | 3778 | |||
XM_011534809.2 | XP_011533111.1 | 19 | 3721 | |||
XM_011534810.2 | XP_011533112.1 | 19 | 3699 | |||
XM_011534812.2 | XP_011533114.1 | 21 | 3731 | |||
GenBank mRNA | AB046765.2 | - | 17 | 3390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
