siRNA Details
Gene Symbol: CUX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015267.3 | NP_056082.2 | 22 | 4651 | ||
XM_011538061.2 | XP_011536363.1 | 22 | 6161 | |||
XM_011538063.2 | XP_011536365.1 | 21 | 6026 | |||
XM_011538069.2 | XP_011536371.1 | 22 | 4500 | |||
XM_011538070.2 | XP_011536372.1 | 22 | 5410 | |||
XM_017019080.1 | XP_016874569.1 | 23 | 5493 | |||
XM_017019081.1 | XP_016874570.1 | 23 | 4556 | |||
GenBank mRNA | AB006631.2 | - | 22 | 4650 | ||
BC151245.1 | - | 22 | 4661 | 8860519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
