siRNA Details
Gene Symbol: MESDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015154.2 | NP_055969.1 | 3 | 560 | ||
NR_126327.1 | 3 | 560 | ||||
GenBank mRNA | AK026606.1 | - | 3 | 488 | ||
AY359110.1 | - | 3 | 546 | |||
BC009210.2 | - | 3 | 510 | 3637449 | ||
BC012746.2 | - | 3 | 510 | 3954629 | ||
D42039.2 | - | 3 | 476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
