siRNA Details
Gene Symbol: MESDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126327.1 | 5 | 2215 | |||
NR_126328.1 | 3 | 1435 | ||||
GenBank mRNA | AK301416.1 | - | 3 | 1355 | ||
BC009210.2 | - | 5 | 2165 | 3637449 | ||
BC012746.2 | - | 5 | 2165 | 3954629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
