siRNA Details
Gene Symbol: TANGO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024562.1 | NP_078838.1 | 4 | 955 | ||
XM_011523327.2 | XP_011521629.1 | 4 | 1014 | |||
GenBank mRNA | AB051533.1 | - | 4 | 955 | ||
AK095333.1 | - | 3 | 501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
