siRNA Details
Gene Symbol: TANGO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024562.1 | NP_078838.1 | 12, 13 | 2139 | ||
XM_011523327.2 | XP_011521629.1 | 12, 13 | 2198 | |||
GenBank mRNA | AB051533.1 | - | 12, 13 | 2139 | ||
AK022750.1 | - | 6, 7 | 803 | |||
AK095333.1 | - | 11, 12 | 1685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
