siRNA Details
Gene Symbol: NDST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301063.1 | NP_001287992.1 | 2 | 926 | ||
NM_001543.4 | NP_001534.1 | 2 | 839 | |||
XM_005268433.1 | XP_005268490.1 | 2 | 811 | |||
XM_005268434.2 | XP_005268491.1 | 2 | 1420 | |||
XM_005268435.4 | XP_005268492.1 | 2 | 945 | |||
XM_005268436.3 | XP_005268493.1 | 2 | 2585 | |||
XM_005268437.4 | XP_005268494.1 | 2 | 824 | |||
XM_005268438.1 | XP_005268495.1 | 2 | 811 | |||
XM_005268439.1 | XP_005268496.1 | 2 | 811 | |||
XM_005268441.1 | XP_005268498.1 | 2 | 811 | |||
XM_006714782.1 | XP_006714845.1 | 2 | 846 | |||
XM_006714783.1 | XP_006714846.1 | 2 | 811 | |||
XM_017009427.1 | XP_016864916.1 | 2 | 3802 | |||
XM_017009428.1 | XP_016864917.1 | 2 | 944 | |||
XM_017009429.1 | XP_016864918.1 | 2 | 824 | |||
XM_017009430.1 | XP_016864919.1 | 2 | 846 | |||
XM_017009431.1 | XP_016864920.1 | 2 | 1416 | |||
GenBank mRNA | AB209107.1 | - | 2 | 749 | ||
AK292448.1 | - | 2 | 839 | |||
AK293746.1 | - | 2 | 926 | |||
BC012888.1 | - | 1 | 560 | 3882074 | ||
U17970.1 | - | 1 | 565 | |||
U18918.1 | - | 1 | 553 | |||
U36600.1 | - | 2 | 760 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
