siRNA Details
Gene Symbol: HMGXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014983.2 | NP_055798.2 | 12 | 2789 | ||
XM_006714765.2 | XP_006714828.1 | 11 | 2291 | |||
GenBank mRNA | BC005880.2 | - | 6 | 793 | ||
BC051025.1 | - | 12 | 2655 | |||
BC092520.1 | - | 12 | 2662 | |||
D83778.1 | - | 12 | 2644 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
