siRNA Details
Gene Symbol: HMGXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014983.2 | NP_055798.2 | 18 | 3715 | ||
XM_006714765.2 | XP_006714828.1 | 17 | 3217 | |||
GenBank mRNA | BC005880.2 | - | 12 | 1719 | ||
BC051025.1 | - | 18 | 3581 | |||
BC063691.1 | - | 4 | 483 | |||
BC092520.1 | - | 18 | 3588 | |||
D83778.1 | - | 18 | 3570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
