siRNA Details
Gene Symbol: FAM214A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286495.1 | NP_001273424.1 | 3, 4 | 578 | ||
NM_019600.3 | NP_062546.2 | 4, 5 | 577 | |||
NR_104457.1 | 4, 5 | 719 | ||||
XM_005254547.2 | XP_005254604.1 | 4, 5 | 540 | |||
XM_005254548.2 | XP_005254605.1 | 4, 5 | 1059 | |||
XM_011521789.2 | XP_011520091.1 | 4, 5 | 606 | |||
XM_011521790.1 | XP_011520092.1 | 3, 4 | 578 | |||
GenBank mRNA | AB037791.1 | - | 4, 5 | 555 | ||
AK292917.1 | - | 4, 5 | 566 | |||
AK293920.1 | - | 3, 4 | 568 | |||
AK308049.1 | - | 3, 4 | 383 | |||
BC040548.1 | - | 4, 5 | 719 | 5288206 | ||
BC109127.1 | - | 3, 4 | 508 | 40036501 | ||
BC109128.2 | - | 3, 4 | 508 | 40036502 | ||
DA101547.1 | - | 3, 4 | 578 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
