siRNA Details
Gene Symbol: THSD7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015204.2 | NP_056019.1 | 5 | 1744 | ||
XM_006715659.1 | XP_006715722.1 | 5 | 1743 | |||
XM_006715660.1 | XP_006715723.1 | 5 | 1743 | |||
XM_006715662.1 | XP_006715725.1 | 5 | 1743 | |||
XM_011515193.2 | XP_011513495.1 | 5 | 4068 | |||
XM_011515194.1 | XP_011513496.1 | 5 | 1509 | |||
GenBank mRNA | AB023177.2 | - | 4 | 1028 | ||
AK092252.1 | - | 5 | 1744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
