siRNA Details
Gene Symbol: RPGRIP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255867.1 | XP_005255924.1 | 17 | 2549 | ||
XM_005255868.1 | XP_005255925.1 | 18 | 2561 | |||
XM_011522970.1 | XP_011521272.1 | 18 | 2561 | |||
XM_011522971.2 | XP_011521273.1 | 18 | 2561 | |||
XM_011522973.2 | XP_011521275.1 | 18 | 2561 | |||
XM_017023094.1 | XP_016878583.1 | 18 | 2561 | |||
XM_017023095.1 | XP_016878584.1 | 18 | 2561 | |||
XM_017023096.1 | XP_016878585.1 | 18 | 2561 | |||
XM_017023098.1 | XP_016878587.1 | 4 | 1524 | |||
XM_017023099.1 | XP_016878588.1 | 4 | 890 | |||
GenBank mRNA | AB023222.1 | - | 13 | 1978 | ||
AK026552.1 | - | 3 | 508 | |||
AK096333.1 | - | 17 | 2560 | |||
AK304790.1 | - | 17 | 2611 | |||
AK307929.1 | - | 17 | 2569 | |||
AK308390.1 | - | 17 | 2560 | |||
BC136433.1 | - | 17 | 2554 | 9020422 | ||
CR749645.1 | - | 17 | 2568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
