siRNA Details
Gene Symbol: RPGRIP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255867.1 | XP_005255924.1 | 23 | 3401 | ||
XM_005255868.1 | XP_005255925.1 | 24 | 3377 | |||
XM_017023094.1 | XP_016878583.1 | 25 | 3515 | |||
XM_017023095.1 | XP_016878584.1 | 23 | 3275 | |||
XM_017023096.1 | XP_016878585.1 | 24 | 3377 | |||
XM_017023098.1 | XP_016878587.1 | 11 | 2478 | |||
XM_017023099.1 | XP_016878588.1 | 11 | 1844 | |||
GenBank mRNA | AB023222.1 | - | 18 | 2692 | ||
AK026552.1 | - | 9 | 1324 | |||
AK304790.1 | - | 23 | 3427 | |||
AK307929.1 | - | 23 | 3421 | |||
BC136433.1 | - | 23 | 3370 | 9020422 | ||
CR749645.1 | - | 24 | 3522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
