siRNA Details
Gene Symbol: RPGRIP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255867.1 | XP_005255924.1 | 26 | 4460 | ||
XM_005255868.1 | XP_005255925.1 | 27 | 4436 | |||
XM_017023094.1 | XP_016878583.1 | 28 | 4574 | |||
XM_017023095.1 | XP_016878584.1 | 26 | 4334 | |||
XM_017023098.1 | XP_016878587.1 | 14 | 3537 | |||
XM_017023099.1 | XP_016878588.1 | 14 | 2903 | |||
GenBank mRNA | AB023222.1 | - | 21 | 3751 | ||
AK026552.1 | - | 12 | 2383 | |||
CR749645.1 | - | 27 | 4581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
