siRNA Details
Gene Symbol: WDR59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324171.1 | NP_001311100.1 | 8 | 679 | ||
NM_001324172.1 | NP_001311101.1 | 8 | 679 | |||
NM_030581.3 | NP_085058.3 | 8 | 652 | |||
XM_005256146.3 | XP_005256203.1 | 8 | 656 | |||
XM_011523331.2 | XP_011521633.1 | 7 | 496 | |||
XM_011523332.2 | XP_011521634.1 | 2 | 79 | |||
XM_017023667.1 | XP_016879156.1 | 7 | 513 | |||
XM_017023668.1 | XP_016879157.1 | 6 | 289 | |||
XM_017023669.1 | XP_016879158.1 | 7 | 384 | |||
XM_017023670.1 | XP_016879159.1 | 6 | 392 | |||
GenBank mRNA | AB067510.1 | - | 3 | 125 | ||
AK302557.1 | - | 8 | 720 | |||
BC004519.2 | - | 8 | 616 | 3938206 | ||
BC014887.2 | - | 2 | 44 | |||
BU850134.1 | - | 8 | 604 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
