siRNA Details
Gene Symbol: WDR59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324171.1 | NP_001311100.1 | 10 | 877 | ||
NM_001324172.1 | NP_001311101.1 | 10 | 877 | |||
NM_030581.3 | NP_085058.3 | 10 | 850 | |||
XM_005256146.3 | XP_005256203.1 | 10 | 854 | |||
XM_011523331.2 | XP_011521633.1 | 9 | 694 | |||
XM_011523332.2 | XP_011521634.1 | 4 | 277 | |||
XM_017023667.1 | XP_016879156.1 | 9 | 711 | |||
XM_017023668.1 | XP_016879157.1 | 8 | 487 | |||
XM_017023669.1 | XP_016879158.1 | 9 | 582 | |||
XM_017023670.1 | XP_016879159.1 | 8 | 590 | |||
GenBank mRNA | AB067510.1 | - | 5 | 323 | ||
AK302557.1 | - | 10 | 918 | |||
BC004519.2 | - | 10 | 814 | 3938206 | ||
BC014887.2 | - | 4 | 242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
