siRNA Details
Gene Symbol: GINS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021067.4 | NP_066545.3 | 4 | 444 | ||
NR_134574.1 | 4 | 444 | ||||
XM_017028162.1 | XP_016883651.1 | 4 | 401 | |||
GenBank mRNA | AK294593.1 | - | 4 | 398 | ||
BC012542.1 | - | 4 | 445 | 4333095 | ||
D80008.1 | - | 4 | 400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
