siRNA Details
Gene Symbol: ISM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080826.1 | NP_543016.1 | 6 | 2457 | ||
GenBank mRNA | BC017997.1 | - | 1 | 1513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
