siRNA Details
Gene Symbol: ZNF629 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080417.1 | NP_001073886.1 | 3 | 2047 | ||
XM_005255222.3 | XP_005255279.1 | 3 | 2186 | |||
GenBank mRNA | AB002324.1 | - | 3 | 2013 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
