siRNA Details
Gene Symbol: GSDMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166237.1 | NP_001159709.1 | 14 | 2302 | ||
NM_024736.6 | NP_079012.3 | 11 | 1748 | |||
XM_011517301.1 | XP_011515603.1 | 10 | 2350 | |||
GenBank mRNA | AF318341.1 | - | 12 | 1787 | ||
AK022212.1 | - | 11 | 1707 | |||
AK093858.1 | - | 5 | 2056 | |||
AK096216.1 | - | 11 | 1495 | |||
AK127941.1 | - | 6 | 3350 | |||
AY008304.1 | - | 11 | 1736 | |||
BC008904.2 | - | 11 | 1733 | 3634992 | ||
BC069000.1 | - | 11 | 1688 | 6213121 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
