siRNA Details
Gene Symbol: GSDMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166237.1 | NP_001159709.1 | 13 | 1856 | ||
NM_024736.6 | NP_079012.3 | 10 | 1302 | |||
XM_011517301.1 | XP_011515603.1 | 9 | 1904 | |||
GenBank mRNA | AF318341.1 | - | 11 | 1340 | ||
AK022212.1 | - | 10 | 1261 | |||
AK093858.1 | - | 4 | 1610 | |||
AK096216.1 | - | 10 | 1049 | |||
AK123985.1 | - | 8 | 1750 | |||
AK127941.1 | - | 5 | 2904 | |||
AY008304.1 | - | 10 | 1290 | |||
BC008904.2 | - | 10 | 1287 | 3634992 | ||
BC069000.1 | - | 10 | 1242 | 6213121 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
