siRNA Details
Gene Symbol: PARD6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032521.2 | NP_115910.1 | 3 | 1079 | ||
GenBank mRNA | AB044555.1 | - | 3 | 836 | ||
BC060847.1 | - | 3 | 1079 | 30345080 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
