siRNA Details
Gene Symbol: OVOL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302757.1 | NP_001289686.1 | 4 | 434 | ||
XM_011527251.2 | XP_011525553.1 | 3 | 414 | |||
XM_017027190.1 | XP_016882679.1 | 3 | 526 | |||
XM_017027191.1 | XP_016882680.1 | 4 | 454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
