siRNA Details
Gene Symbol: TTC39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080494.3 | NP_001073963.1 | 3 | 270 | ||
NM_001144832.2 | NP_001138304.1 | 3 | 245 | |||
NM_001297662.1 | NP_001284591.1 | 3 | 247 | |||
NM_001297663.1 | NP_001284592.1 | 3 | 247 | |||
NM_001297664.1 | NP_001284593.1 | 3 | 283 | |||
NM_001297665.1 | NP_001284594.1 | 3 | 270 | |||
XM_005270644.2 | XP_005270701.1 | 3 | 276 | |||
XM_006710471.2 | XP_006710534.1 | 3 | 268 | |||
XM_011541048.2 | XP_011539350.1 | 3 | 245 | |||
XM_011541049.2 | XP_011539351.1 | 3 | 384 | |||
XM_011541050.2 | XP_011539352.1 | 3 | 245 | |||
XM_011541051.2 | XP_011539353.1 | 3 | 278 | |||
XM_011541052.2 | XP_011539354.1 | 3 | 224 | |||
XM_011541053.2 | XP_011539355.1 | 3 | 262 | |||
XM_011541054.2 | XP_011539356.1 | 3 | 203 | |||
XM_017000715.1 | XP_016856204.1 | 3 | 250 | |||
GenBank mRNA | AB007921.1 | - | 3 | 293 | ||
AF007170.1 | - | 3 | 151 | |||
AK000220.1 | - | 3 | 245 | |||
AK298019.1 | - | 3 | 254 | |||
AK301587.1 | - | 3 | 243 | |||
AK311430.1 | - | 3 | 172 | |||
BI761577.1 | - | 3 | 283 | |||
CB128766.1 | - | 3 | 247 | |||
CX788693.1 | - | 3 | 185 | |||
DA871604.1 | - | 3 | 245 | |||
DB460455.1 | - | 3 | 268 | |||
DC381029.1 | - | 3 | 242 | |||
HY029911.1 | - | 3 | 390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
