siRNA Details
Gene Symbol: SHANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033517.1 | NP_277052.1 | 22 | 6855 | ||
GenBank mRNA | AB051437.1 | - | 2 | 4054 | ||
AK074038.1 | - | 2 | 3974 | |||
BC018856.2 | - | 1 | 919 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
