siRNA Details
Gene Symbol: MAST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015016.1 | NP_055831.1 | 12 | 1095 | ||
XM_005259823.3 | XP_005259880.1 | 12 | 1459 | |||
XM_005259824.2 | XP_005259881.1 | 13 | 1145 | |||
XM_005259825.2 | XP_005259882.1 | 13 | 1142 | |||
XM_005259826.3 | XP_005259883.1 | 11 | 1255 | |||
XM_005259827.2 | XP_005259884.1 | 11 | 1113 | |||
XM_006722698.2 | XP_006722761.1 | 12 | 1137 | |||
XM_006722699.2 | XP_006722762.2 | 12 | 1446 | |||
XM_006722700.3 | XP_006722763.1 | 11 | 1255 | |||
XM_011527823.1 | XP_011526125.1 | 14 | 1235 | |||
XM_017026506.1 | XP_016881995.1 | 12 | 1443 | |||
XM_017026507.1 | XP_016881996.1 | 11 | 1425 | |||
XM_017026508.1 | XP_016881997.1 | 11 | 1423 | |||
XM_017026509.1 | XP_016881998.1 | 12 | 1446 | |||
XM_017026510.1 | XP_016881999.1 | 11 | 1426 | |||
XM_017026511.1 | XP_016882000.1 | 13 | 1533 | |||
XM_017026512.1 | XP_016882001.1 | 12 | 1124 | |||
XM_017026513.1 | XP_016882002.1 | 12 | 1121 | |||
XM_017026514.1 | XP_016882003.1 | 13 | 1128 | |||
XM_017026515.1 | XP_016882004.1 | 12 | 1134 | |||
XM_017026516.1 | XP_016882005.1 | 12 | 1443 | |||
GenBank mRNA | AB011133.1 | - | 12 | 1093 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
