siRNA Details
Gene Symbol: MAST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015016.1 | NP_055831.1 | 27 | 3973 | ||
XM_005259823.3 | XP_005259880.1 | 27 | 4364 | |||
XM_005259824.2 | XP_005259881.1 | 28 | 4050 | |||
XM_005259825.2 | XP_005259882.1 | 28 | 4047 | |||
XM_005259826.3 | XP_005259883.1 | 26 | 4160 | |||
XM_006722698.2 | XP_006722761.1 | 27 | 4042 | |||
XM_006722699.2 | XP_006722762.2 | 27 | 4351 | |||
XM_006722700.3 | XP_006722763.1 | 26 | 4133 | |||
XM_011527823.1 | XP_011526125.1 | 29 | 4140 | |||
XM_017026506.1 | XP_016881995.1 | 27 | 4348 | |||
XM_017026507.1 | XP_016881996.1 | 26 | 4330 | |||
XM_017026508.1 | XP_016881997.1 | 26 | 4328 | |||
XM_017026509.1 | XP_016881998.1 | 27 | 4324 | |||
XM_017026511.1 | XP_016882000.1 | 28 | 4438 | |||
XM_017026512.1 | XP_016882001.1 | 27 | 4029 | |||
XM_017026513.1 | XP_016882002.1 | 27 | 4026 | |||
XM_017026514.1 | XP_016882003.1 | 28 | 4033 | |||
XM_017026515.1 | XP_016882004.1 | 27 | 4039 | |||
XM_017026516.1 | XP_016882005.1 | 27 | 4321 | |||
GenBank mRNA | AB011133.1 | - | 27 | 3971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
