siRNA Details
Gene Symbol: ERI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301698.1 | NP_001288627.1 | 2 | 414 | ||
NM_001301699.1 | NP_001288628.1 | 2 | 46 | |||
NM_001301700.1 | NP_001288629.1 | 3 | 122 | |||
NM_024066.2 | NP_076971.1 | 3 | 490 | |||
XM_005271186.2 | XP_005271243.1 | 3 | 478 | |||
XM_017002301.1 | XP_016857790.1 | 3 | 472 | |||
XM_017002302.1 | XP_016857791.1 | 1 | 39 | |||
XM_017002303.1 | XP_016857792.1 | 3 | 121 | |||
GenBank mRNA | AF007157.1 | - | 3 | 419 | ||
AK290567.1 | - | 3 | 478 | |||
AK297708.1 | - | 1 | 36 | |||
BC144384.1 | - | 2 | 388 | |||
BG323839.1 | - | 3 | 458 | |||
BI765887.1 | - | 2 | 118 | |||
DC413416.1 | - | 3 | 122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
